Learn More
DDX5 Mouse anti-Human, CoraLite 488, Clone: 2D3D2, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc CL48867025100UL
This item is not returnable.
View return policy
Description
The DDX5 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human DDX5. This antibody recognizes human, mouse, rat antigen. The DDX5 antibody has been validated for the following applications: FC, IF analysis. This antibody is coralite™488.
Specifications
DDX5 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Mouse | |
IgG1 |
Immunofluorescence, Flow Cytometry | |
2D3D2 | |
CoraLite 488 | |
100 μL | |
1655 | |
Liquid |