Learn More
MOUSER ELECTRONICS DC FANS DC FAN, 80X80X15MM, 12
Supplier:  MOUSER ELECTRONICS 369ME80151V1000UA99
Encompass
DC Fans DC Fan,80x80x15mm, 12VDC,37CFM, 1.96W,34.7dBA, 3000RPM,Vapo, Auto RestartRoHS Compliant
Catalog No. NC2235918