Learn More
Crescent Chemical Co Inc CYCLOHEXIMIDE CRYST. PURE
Supplier: Crescent Chemical Co Inc SE10700.02
Cycloheximide cryst. pure Glutarimide antibiotic isolated from Streptomyces griseus; active against fungi and yeasts, but not against bacteria. Inhibits eukaryotic protein synthesis by blocking the translocation step in the elongation cycle (1,3,8). Blocks tra