Learn More
Revvity Health Sciences Inc Cyanine 3-CTP, 100 nmol
Supplier: Revvity Health Sciences Inc NEL580001EA
NEN brand fluorescent nucleotide analogs are biologically active with a variety of DNA and/or RNA polymerases. Labeling methods such as: nick translation, random priming, polymerase chain reaction, 3'-end labeling, or transcription of RNA using SP6, T3, or T7 RNA polymerases may be used. Some analogs demonstrate variations in relative performance depending upon nucleotide and fluorophore selected due to enzyme preferences. Labeled probes may be used in applications including (but not limited to) microarrays and chromosome mapping. These analogs are intended to be detected directly by their fluorescence properties.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.