Learn More
Advanced ImmunoChemical, Inc. Recombinant Human Insulin-Like Growth Factor Binding Protein 4, C-Terminal Fragment (CT-IGFBP- 4), Source - Expressed In Human Cell Line. Purity > 95% (TRICINE-SDS-PAGE), 0.05mg
Supplier:  Advanced ImmunoChemical, Inc. 8IGBPCRH0.05MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Antigens / cardiac markers / Recombinant human Insulin-like growth factor binding protein 4, C-terminal fragment (CT-IGFBP- 4). Source:Expressed in human cell line. CT-IGFBP-4 is expressed as a single polypeptide chain (Lys157-Glu258, UniProt ID = P22692) without tags.Analysis:Purity > 95% (Tricine-SDS-PAGE).CT-IGFBP4 concentration is determined spectrophotometrically.Immunoreactivity is confirmed by reaction with monoclonal antibodies specific to different epitopes of CT-IGFBP-4 molecule.Purification:Immunoaffinity purificationPresentation:Lyophilized from 20 mM Tris-HCl, 150 mM NaCl, pH 7.4Storage Conditions:Before and after reconstitution at -70 °C (between -65 and -80 °C allowed)Other information:Avoid repeated freezing and thawing.It is recommended to aliquot the product after the first thawing. This product is sold for research use only.
Catalog No. 50-153-9842