Learn More
LECO Corporation Ceramic Combustion Boats, 100 per pack
Supplier: LECO Corporation 529203
Unglazed, Round Bottom Shape. For macrosamples/horizontal-loading furnaces. For use with 632 Series, 628 Series Module, TruSpec Series Module, TruMac Series.