Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Color Standard, APHA/Hazen (Platinum-Cobalt), Color = 50, Ricca Chemical
$119.30 - $539.50
Specifications
Chemical Name or Material | Color Standard |
---|---|
Name Note | Color = 50 |
Packaging | Amber Glass Bottle |
CAS Min % | 97.0, 0.96, 0.01, 0.01 |
CAS Max % | 100.0, 0.99, 0.01, 0.01 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
R2230050100
|
Ricca Chemical Company
2230.050100 |
100 mL | Amber Glass Bottle |
Each for $119.30
|
|
||||
2230050-16
|
Ricca Chemical Company
2230.05016 |
500 mL | Amber Glass Bottle |
Each for $321.50
|
|
||||
R22300504C
|
Ricca Chemical Company
2230.0501 |
4 L | Amber Glass Bottle |
Each for $539.50
|
|
||||
Specifications
Color Standard | |
Amber Glass Bottle | |
100.0, 0.99, 0.01, 0.01 | |
Brown/Yellow to Yellow | |
Liquid |
Color = 50 | |
97.0, 0.96, 0.01, 0.01 | |
7647-01-0, 16921-30-5, 7791-13-1, 7647-01-0, 7732-18-5 | |
APHA / Hazen (Platinum-Cobalt) |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title