Learn More
Thermo Scientific Chemicals Collagenase, Type I, Clostridium histolyticum
Type I Collagenase (CAS # 9001-12-1) is one of seven collagenases, enzymes responsible for collagen degradation.
$95.50 - $431.50
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AAJ62406MC
|
Thermo Scientific Chemicals
J62406MC |
100 mg |
Each for $95.50
Order in Bulk
Request bulk or custom formats |
|
|||||
AAJ6240603
|
Thermo Scientific Chemicals
J6240603 |
1 g |
Each for $431.50
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
Collagenase, Type I, Clostridium histolyticum is used as a powder and solvent injection kit for the treatment of dupuytren's contracture.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
- Type I collagenase is a collagen-specific enzyme required for the proper degradation of connective tissue
- Type I collagenase is isolated from Clostridium histolyticum
- It is implicated in connective tissue disorders such as Peyronie’s and Dupuytren’s
Application
- Type I collagenase targets specific alpha helices within collagen
- It is suitable for use in cell culture and biochemistry applications
Specifications
Powder |
Safety and Handling
EINECSNumber : 232-582-9
Recommended Storage : Keep cold
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.