Learn More
CHEMSCIENCE Volumetric Flask, 2000mL, Serialized and Certified, Heavy Duty, Wide Mouth, PE Stopper# 27, Class A,PK/2
Supplier:  CHEMSCIENCE 129.503.10
Encompass_Preferred
Wide mouth design facilitates mixing, pipette access, sampling and filling. Unique flat bottom interior permits the use of a standard stir bar. The graduation line on the neck is sharp with permanent large colored encryption for easy-read. Flasks are calibrated 'To Contain' at Class A tolerances, per ASTM E288 specifications. Supplied with closed bottom Polyethylene stopper to fit in the ground neck.
Catalog No. NC2681273