Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Choice™ Polypropylene (PP) Syringe Filters

Use Choice Polypropylene Syringe Filters as an economic solution to filtration of biological samples, and for filtration of aggressive organic solutions.

Supplier:  Thermo Scientific CH2225PP

 View more versions of this product

Catalog No. 03-253-904


Add to Cart

Description

Description

Thermo Scientific™ Choice™ Polypropylene Syringe Filters are a good choice for filtration of biological samples, and for filtration of aggressive organic solutions. They provide cleaner sample extracts by removing interfering materials and fine particles down to the rated pore size.

Adding a syringe filtration step prior to chromatographic injection not only helps to ensure more consistent and reliable results, it also helps protect instruments and prolong column life. Economic Choice Syringe Filters are available in a variety of sizes and membrane types to meet any application need.

Specifications
  • Membrane: Hydrophilic polypropylene
  • Maximum operating temperature: 110 °C
  • Housing: Medical grade, virgin polypropylene
Applications
  • Protein- or peptide-based assays
  • General HPLC analysis
Chemical Incompatibilities
  • Hexane, toluene, benzene, limited resistance to dichloromethane and chloroform
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Choice Polypropylene (PP) Syringe Filters 0.2 μm, 25 mm, 100 Pk.
Polypropylene
0.2 μm
HPLC, SPE
100
90 psi
Syringe Filter
Single-use
Polypropylene
Non-sterile
Solvent Resistent, Low-Extractable Polypropylene
No
Choice
25 mm
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit