Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eppendorf™ Centrifuge 5910 Ri PROMO

The refrigerated Centrifuge 5910 Ri combines high capacity and performance in a compact footprint with an ergonomic design. An intuitive VisioNize touch interface, advanced temperature management, and full portfolio of rotors make this centrifuge ideal for a broad range of applications.

Supplier:  Eppendorf™ 2231001018

Request Bulk Order

Catalog No. 05-414-459PM


Add to Cart

Description

Description

  • Large 7-inch VisioNize touch interface for super-fast parameter settings via unique favorites function
  • Swing-bucket rotors and adapters for tubes and bottles from 0.2 mL to 1000 mL, as well as all types of MTP, PCR, or Deepwell Plates
  • Fixed-angle rotors for high-speed molecular biology applications in tubes from 0.2 mL to 250 mL
  • High centrifugation speed of up to 22,132 x g (14,000 rpm)
  • User management with multiple authorization levels and documentation features supporting GLP/GxP requirements
  • Aerosol-tight Eppendorf QuickLock caps and lids for fast and ergonomic lid locking
  • Quiet operation to improve your work environment
  • Compact footprint to save valuable bench space
  • Automatic rotor recognition and imbalance detection for maximum operational safety
TRUSTED_SUSTAINABILITY
Specifications

Specifications

4 x 1000 mL
14,000 rpm (4500 with included rotor)
VisioNize™ Touch Interface
99 Programs
Yes
10 sec. to 99 hr. 59 min.
68 x 72 x 37 cm (28.3 x 25.9 x 14.5 in.)
85 cm
120 V
50/60 Hz
22,132 x g (4,347 x g with included rotor)
Yes
10
S-4xUniversal
-11°C to +40°C
5910 Ri with Rotor
33.5 in.
109 kg
12 A
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit