Learn More
Abnova™ CEN17 CISH Probe
CEN17 CISH Probe is designed for the qualitative detection of human chromosome 17 alpha satellites in formalin-fixed, paraffin-embedded specimens by chromogenic in situ hybridization (CISH).
Supplier: Abnova™ CC0001
Specifications
Reagent Provided: 1. Digoxigenin-labeled polynucleotides targeting sequences mapping in 17p11.1-q11.1 specific for the alpha satellite centromeric region D17Z1 of chromosome 17 2. Formamide based hybridization buffer |
|
The probe is intended to be used in combination with the CISH Implementation Kit 1, which provides necessary reagents for specimen pretreatment and post-hybridization processing. | |
The product is ready-to-use. No reconstitution, mixing, or dilution is required. Bring probe to room temperature (18 to 25°C) and mix briefly before use. |