Learn More
Thermo Scientific™ CaptureSelect™ Transferrin Affinity Matrix
This is one of a group of CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins.
$1178.00 - $10300.00
Specifications
Form | Liquid Suspension |
---|---|
Product Line | CaptureSelect™ |
Type | Affinity Matrix |
Stationary Phase | Anti-transferrin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
19-130-6005
|
Thermo Scientific™
191306005 |
5 mL |
Each for $1,178.00
|
|
|||||
19-130-6010
|
Thermo Scientific™
191306010 |
10 mL |
Each for $2,170.00
|
|
|||||
19-130-6050
|
Thermo Scientific™
191306050 |
50 mL |
Each for $10,300.00
|
|
|||||
Description
This is one of a group of CaptureSelect™ protein purification affinity resins developed for the simple, single-step purification of a variety human plasma and serum proteins. This unique selection of affinity purification resins can purify proteins or other non-antibody biomolecules from either recombinant or native sources with a high degree of affinity and specificity, and are immobilized on a high capacity agarose support.
Specifications
Liquid Suspension | |
CaptureSelect™ | |
Affinity Matrix | |
Anti-transferrin |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.