Learn More
Enzo Life Sciences SCREEN-WELL Cancer Library (100µl/well)
Supplier: Enzo Life Sciences ENZLIB1020100
The SCREEN-WELL Cancer Library is a collection of 275 compounds that can be used for cancer inhibitor screening and assay development. The library contains small molecules affecting mTOR, aurora kinases, tyrosine kinases, PI3K, and HDAC, as well as many structurally and mechanistically different compound classes. This library is an essential tool for cancer inhibitor screening and drug development. The compounds are dissolved in DMSO at 10 mM and aliquoted into 96-well deep-well plates at 100 µL per well. Long Term Storage: -80°C. Applications: HTS .
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.