Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Estes Industries, LLC C11-3 Engines
The Estes C11-3 engine is a single stage engine designed for model rocket flight. This engine is a 24mm diameter engine designed for rockets with a maximum lift weight of less than 6 ounces. Each package includes 2 engines, 2 starters and 4 starter plugs.
Supplier: Estes Industries, LLC 001522
Description
- The Estes C11-3 engine is a single stage engine designed for model rocket flight.
- This engine is a 24mm diameter engine designed for rockets with a maximum lift weight of less than 6 ounces.
- Each package includes 2 engines
Specifications
Rockets | |
Propellant | |
2 engines, 2 starters and 4 starter plugs. | |
0.94 in. | |
1.13 oz. |
Propellant | |
Model Rocket | |
7 in. | |
2.75 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction