Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Brady™ Key Retaining Keyed Different Steel Padlocks with 3 inch Shackle Locks

Key retaining safety feature prevents key from being removed until closed

Supplier:  Brady™ 123236

 View more versions of this product

Catalog No. 19-103-383

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Includes: 1 key

Description

Description

  • Key-retaining steel padlock has a 3 in. reinforced laminated steel shackle
  • Keyed different
  • Hardened steel body withstands severe physical abuse and tampering
  • Offers a lighterweight option in comparison to standard laminated steel padlocks
  • Available in a variety of colors
Specifications

Specifications

Red
Product Suggestions

Product Suggestions

Safety and Handling

Safety and Handling

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit