Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sartorius Bottled Media for Sartorius Membrane Filtration Units
Lysine agar media for wild yeasts determination
Specifications
Form | Liquid |
---|---|
Manufacturing Origin | Germany |
Fill Volume | 250 mL |
Packaging Type | Bottle |
Quantity | 250 mL |
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Application) | Price | Quantity & Availability | |||||
5090117907
|
Sartorius
14157A |
Membrane filtration method or for direct incubation, for determination of Yeasts and molds |
Pack of 4 for $123.00
|
N/A | |||||
5090117904
|
Sartorius
14148A |
Membrane filtration method or for direct incubation, for determination of LactoBacilli and Pediococci and other beer spoiling organisms |
Pack of 4 for $169.00
|
N/A | |||||
14-555-740
|
Sartorius
14143A |
Membrane filtration method or for direct incubation, for determination of wild Yeasts | N/A | N/A | N/A | ||||
5090117903
|
Sartorius
14144A |
Membrane filtration method or for direct incubation, for determination of total count | N/A | N/A | N/A | ||||
5090117905
|
Sartorius
14155A |
Membrane filtration method or for direct incubation, for determination of Escherichia coli and Coliforms | N/A | N/A | N/A | ||||
Description
The traditional culture media for microorganisms is agar media. This can be used for the membrane filtration method or for direct incubation. Agar media in bottles are the cost-effective alternative for casting plates.
- 250mL bottles, 4 bottles per box
- Economical, provides time and cost savings
- According to international formulas
- Consistent quality
Specifications
Liquid | |
250 mL | |
250 mL |
Germany | |
Bottle |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title