Learn More
Ace Glass, Inc. Bottle only, pressure, 14mL, #7, no plug (Actual fill capacity 14mL)
Supplier: Ace Glass, Inc. 8648124
14mL pressure bottle with #7 Ace-Thred, rated for 60psig at room temperature. Does not include plug.