Learn More
Chemglass Life Sciences Bottle, Hybridization, 35 x 150mm, GL-45, Red Cap
Supplier: Chemglass Life Sciences CG114003
Bottle, Hybridization, 35 x 150mm, GL-45, Red Cap. Heavy wall bottle used in hybridization incubators or ovens with rotators. An alternative to plastic bags or boxes, the membrane is rolled and placed into the bottle which is secured in a rotisserie. The membrane is saturated and drained during each revolution, resulting in even exposure of the blots to probe or wash solutions. Bottle fits most hybridization equipment and has straight interior walls for easy membrane placement and removal, as well as decanting of hybridization solutions. The red plastic cap has a PTFE faced liner and a working temperature of 180°C. The blue cap has a silicone sealing ring and a working temperature of 140°C.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.