Learn More
Enzo Life Sciences Bone morphogenetic protein 2 (human), (recombinant) (10µg)
Supplier: Enzo Life Sciences ENZPRT2000010
Alternative name: BMP-2. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from 1 mg/mL in 1X PBS. Source: Produced in HEK cells. Glycosylated disulfide-linked homodimer. Long Term Storage: -20°C.