Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SW Sustainability Solutions Bantam™ P-021-098-WT White TPE Single-Use Gloves
Click to view available options
Size:
Large
X-Large
Description
- Food safe
- High tensile strength and stretchability
- Industrial use 4.0 AQL
- Outperforms vinyl gloves
- Latex free
- Powder free
- Recyclable
Specifications
Certifications/Compliance (Europe) | EN ISO 21420:2020, EU 1907/2006, EU 2016/425 |
Applications | Industrial |
AQL | 4 |
Color | White |
Disposable | Single-use |
Exterior Finish | No Texture |
Finger Thickness (English) | 2.1 mil |
Finger Thickness (Metric) | 0.05 mm |
Latex Free | Yes |
Material | TPE |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction