Learn More
Kamiya Biomedical Apo AI Reagent, 60mL+20mL/PK RX
Supplier:  Kamiya Biomedical KAI002
Encompass_Preferred
K-ASSAY Apo AI Assay Reagent. R1: 3 x 20mL (liquid), R2: 1 x 20mL (liquid). Assay Range: 20 - 300 mg/dL. The K-ASSAY Apo AI assay is intended for the quantitative determination of human Apolipoprotein AI (Apo AI) in serum by immunoturbidimetric assay. FOR IN VITRO DIAGNOSTIC USE. Required Calibrator: K-ASSAY Apo AI/B Calibrator (Cat. No. KAI-008C). Recommended Controls: K-ASSAY Liquid Lipid Control (Cat. No. K110C-2M). This assay is intended for use on most popular chemistry analyzers that allow open channel reagents. Please contact KAMIYA BIOMEDICAL COMPANY to check parameter availability for your analyzer.
Catalog No. 50-130-5943
Due to product restrictions, please to purchase or view availability for this product.