Learn More
Morwell MD Biosciences ANTIBODY COCKTAIL 50 MG
SDP
Supplier:  Morwell MD Biosciences CIAMAB50
Encompass
ArthritoMab™ Arthritis Inducing Antibody Cocktail is a cocktail of 4 arthritogenic monoclonal antibodies to collagen II (CII) used for inducing arthritis in the anti-Collagen Antibody Induced Arthritis (CAIA) model.
Catalog No. NC2467665