Learn More
PBL Assay Science Anti-Human Interferon Alpha (IFN-Alpha) Mouse Monoclonal Antibody, Clone MMHA-2 (MAb) [50 micrograms (ug)]
Supplier: PBL Assay Science 211001
Mouse monoclonal antibody Anti-Human Interferon Alpha, Clone MMHA-2 is manufactured by PBL Assay Science (Catalog #21100-1) for research use only (RUO) and packaged in 50 micrograms (ug). This MAb exhibits high binding affinity and is suitable for use in neutralization, ELISA, and Western Blot applications.