Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Glycated Hemoglobin, Human, HBA1C, 0.2mg
Supplier:  Advanced ImmunoChemical, Inc. 2GHH0.2MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal Antibodies / Metabolic Syndrome / Monoclonal mouse anti-glycated hemoglobin, human, HbA1c. MAbs: 75C9Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with glycated (in vitro) synthetic peptide corresponding to the N-terminal end of beta-globin. Specificity: Native human HbA1c, does not cross-react with non-glycated hemoglobin. MAb Isotypes:IgG1 for MAb 75C9. Applications: HbA1c immunoassays. Purification: Chromatography on protein A Sepharose. Presentation: PBS, pH 7.4, 0.1 % sodium azide (NaN3). Storage Conditions: + 4 °C. Material Safety Note: This product is sold as an antibody preparation for research use only. Standard Laboratory Practices should be followed when handling this material. Contains sodium azide (0.09 %) as preservative.
Catalog No. 50-153-9838