Learn More
Electron Microscopy Sciences Annular Sputter Target Platinum, 87 mm O.D., 57 mm I.D.
SDP
Supplier:  Electron Microscopy Sciences 91031PT
Encompass_Preferred
Targets have a standard thickness of 0.1 mm (100 micron) or use on most sputtering units and are available mounted or unmounted. Made from high purity metals. When ordering, please state make and model of the sputtering unit to ensure proper fit.
O.D.: 87 mm
I.D.: 57 mm
Catalog No. 50-328-73
Due to product restrictions, please to purchase or view availability for this product.