Learn More
Perkin Elmer US LLC Autosampler Aluminum Sample Pans - 50 µL, Qty. 400
Supplier: Perkin Elmer US LLC B0143017
These volatile pans and covers are suitable for use with DSC autosamplers, but are also suitable for use with non-robotic systems. Most of these pans and covers can be crimped using the Universal Crimper (B0139005).