Learn More
Electron Microscopy Sciences Omniprobe All Tungsten Probe Tip
Supplier: Electron Microscopy Sciences 7596002
Custom tip designed from tungsten. Tip radius is 0.5um with 13 degree taper angle for maximum lifetime service.