Learn More
MilliporeSigma™ All-Glass Filter Holder Kit, 90 mm
Vacuum filter aqueous, organic, or corrosive liquids for particulate contamination analysis.
Supplier: MilliporeSigma™ XX1019020
Description
Vacuum filter aqueous, organic, or corrosive liquids for particulate contamination analysis.
Specifications
Not Autoclavable | |
Borosilicate Glass | |
Glass Filter Holder Kit |
Glass Filter Holder kit with stainless steel screen membrane support | |
Non-sterile | |
90 mm |