Learn More
Across International Ai SolventVap 5.3-Gallon/20L Rotary Evaporator w/ Motorized Lift
SDP
Supplier:  Across International SE53
Encompass_Preferred
The Ai SolventVap series 20L rotary evaporators is now ETL tested to UL/CSA standard. This certification is required by many municipalities, and it also provides a higher standard of equipment safety and lifespan. Also come standard is the new plexglass bath cover. Specially designed for Ai 20L SolventVap rotary evaporators, this beneficial cover securely rests on top of the water bath and ensures the best possible temperature uniformity. SolventVap rotary evaporators come with all PTFE valves for solvent-resistant operations. We also lowered the height of assembled unit, for easy installation and ease of use and storage. Also come with them are vertical glassware and digital temperature/rotation control. Its optimized bath volume allows for rapid heat-up times.
Catalog No. 50-225-1295