Learn More
Across International Ai SolventVap 2.6-Gallon/10L Rotary Evaporator w/ Motorized Lift
SDP
Supplier:  Across International SE26
Encompass_Preferred
Ai SolventVap 10L series rotary evaporatorsis now ETL tested to UL/CSA standards. This certification is required by many municipalities, and it also provides a higher standard of equipment safety and lifespan. SolventVap comes with all PTFE valves for solvent-resistant operations. We also lowered the height of assembled unit, for easy installation and ease of use and storage. Also come with them are vertical glassware and digital temperature/rotation control. Its optimized bath volume allows for rapid heat-up times. The condenser unit has specially designed glass tubes that utilize surface area extremely efficiently. Thats why our SolventVap evaporators consistently deliver reliable and reproducible process results that are usually reserved for more costly systems. Another important aspect of the SolventVap is the safety features, which includes dry-run protection, water bath over-temperature shut-off protection, and fuse protection.
Catalog No. 50-225-1294