Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Accuform Signs Fluorescent Color Tapes
Mark a wide variety of areas and objects with easily seen fluorescent flagging tape.
$7.78 - $8.76
Specifications
Adhesive Type | Non-adhesive |
---|---|
Dimensions (L x W) | 150 ft. x 1.18 in. |
Material | Vinyl |
Product Type | Flagging Tape |
For Use With (Application) | Marking hazards, trees, danger areas, survey markers and boundaries |
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity & Availability | |||||
19-169-574
|
Accuform Signs
PTL410FO |
Fluorescent Orange |
Each for $7.78
|
|
|||||
19-169-573
|
Accuform Signs
PTL410FR |
Fluorescent Red |
Each for $8.76
|
|
|||||
Specifications
Non-adhesive | |
Vinyl | |
Marking hazards, trees, danger areas, survey markers and boundaries | |
45.7 m | |
4 mil |
150 ft. x 1.18 in. | |
Flagging Tape | |
150 ft. | |
1 Ea. | |
1.18 in. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title