Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Accessories for Thermo Scientific™ Tube Revolver
Optimize flexibility and performance
$17.50 - $25.50
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
S08018
|
Thermo Scientific™
88881008/EMD |
Paddles 0.5mL/0.8mL |
Pack of 2 for $18.00
|
|
|||||
S08015
|
Thermo Scientific™
88881005/EMD |
Paddles 10mL/15mL |
Pack of 2 for $17.50
|
|
|||||
S08017
|
Thermo Scientific™
88881007/EMD |
Paddles 1.5mL/2mL |
Pack of 2 for $18.00
|
|
|||||
S08019
|
Thermo Scientific™
88881009/EMD |
Paddles 50mL |
Each for $25.50
|
|
|||||
Description
Specifications
Tube Revolver |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title