Learn More
Bioassay Systems Parallel Artificial Membrane Permeability Assay (PAMPA) Kit. For assess gastrointestinal (GI) membrane permeability of test compounds.
Supplier: Bioassay Systems PAMPA096
For quantitative determination of gastrointestinal (GI) membrane permeability of test compounds. Key Features: Convenient. Includes all necessary equipment to run a PAMPA plate. Simple and low-cost. Procedure is easy to follow and more affordable than cell-based permeability assays. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: PAMPA. Samples: Chemical compounds. Species: All. Procedure: Assay takes 20 hr, hands-on time 1 hr. Kit size: 96 tests.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.