Learn More
Chemglass Life Sciences 900mL Style Freeze Dry Flask, P/N 7542900
Supplier: Chemglass Life Sciences CG161107
900mL Style Freeze Dry Flask, P/N 7542900. Flat bottom borosilicate flask for use with and Virtis freeze dryers. Wide mouth design provides easy access sample.