Learn More
Chemglass Life Sciences Rushton Style Impeller Blade for 15L Bio Reactors, 76mm Dia, 15mm Shaft Size
SDP
Supplier:  Chemglass Life Sciences CLS138520
Encompass
Rushton Style Impeller Blade for 15L Bio Reactors, 76mm Dia, 15mm Shaft Size. The 3 through 7 Liter sizes fit the 8mm Shaft, while the 15 liter fits the 15mm shaft. These blades are used primarily for bacterial cell culture work.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-129-4461