Learn More
Chemglass Life Sciences 6000mL Glassblowers Erlenmeyer Blank
Supplier: Chemglass Life Sciences CG62010
6000mL Glassblowers Erlenmeyer Blank. Blanks for fabrication of Erlenmeyer shaped flasks. NOTE: NECK IS UNFINISHED, FLASK CANNOT BE USED WITHOUT MODIFICATION OF NECK.