Learn More
Ace Glass, Inc. 250mL 24/40 cylinder. With full length standard taper joints
Supplier: Ace Glass, Inc. 619521
250mL 24/40 cylinder. With full length standard taper joints, single scale metric graduations. Can be used for distilling receivers.