Learn More
Bioassay Systems QuantiFluo™ DNA Assay Kit. For quantitative determination of DNA. For quantitative determination of DNA.
Supplier: Bioassay Systems QFDN250
For quantitative determination of DNA. Key Features: Sensitive and accurate. Linear detection range 2 ng to 40 ng (100 - 2,000 ng/mL) calf thymus DNA in 96-well plate assay. Simple and high-throughput. The "mix-and-read" procedure involves addition of a single working reagent and reading the fluorescence intensity. Can be readily automated as a high-throughput assay for thousands of samples per day. Low interference. RNA, salt (up to 3M NaCl), detergent (< 0.01% SDS) and common DNA extraction buffer do not interfere in the assay. Method: FL350/450nm. Samples: Plasmid DNA, genomic DNA, cDNA, DNA following polymerase chain reaction, and DNA extracted from gel and other matrices. Species: All. Procedure: Assay takes 5 min. Kit size: 250 tests. Detection limit: 100 ng/mL.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.