Learn More
Chemglass Life Sciences 2000ml Glassblowers Heavy Wall Filter Flask Blank
SDP
Supplier:  Chemglass Life Sciences CG62304
Encompass_Preferred
2000ml Glassblowers Heavy Wall Filter Flask Blank. Extra heavy, unmarked blanks for fabrication of filter apparatus. Top has a tooled finish to accept a rubber stopper or neoprene filter adapter.
Catalog No. CG62304