Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,2-Trifluoroethyl acetate, 97%, Thermo Scientific Chemicals

Catalog Number AAL1687114
Click to view available options
Quantity:
25 g
406-95-1
C4H5F3O2
142.08
MFCD00198018
ZOWSJJBOQDKOHI-UHFFFAOYSA-N
trifluoroethyl acetate, unii-en07rvl78u, en07rvl78u, acetic acid, 2,2,2-trifluoroethyl ester, acetic acid 2,2,2-trifluoroethyl ester, trifluoroethylacetate, 2,2,2-trifluoroethylacetate, acmc-20aokx, cf3ch2o2cch3, ksc494s2j
136256
2,2,2-trifluoroethyl acetate
CC(=O)OCC(F)(F)F
406-95-1
C4H5F3O2
142.08
MFCD00198018
ZOWSJJBOQDKOHI-UHFFFAOYSA-N
trifluoroethyl acetate, unii-en07rvl78u, en07rvl78u, acetic acid, 2,2,2-trifluoroethyl ester, acetic acid 2,2,2-trifluoroethyl ester, trifluoroethylacetate, 2,2,2-trifluoroethylacetate, acmc-20aokx, cf3ch2o2cch3, ksc494s2j
136256
2,2,2-trifluoroethyl acetate
CC(=O)OCC(F)(F)F

2,2,2-Trifluoroethyl trifluoroacetate is used in the preparation of dimethyl (3,3,3-trifluoro-2,2-dihydroxypropyl)phosphonate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trifluoroethyl trifluoroacetate is used in the preparation of dimethyl (3,3,3-trifluoro-2,2-dihydroxypropyl)phosphonate

Solubility
Soluble in water (16 g/L) (25°C)

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Density 1.258
Boiling Point 78°C
Flash Point 11°C (51°F)
Refractive Index 1.32
Linear Formula CH3CO2CH2CF3
Quantity 25 g
UN Number UN3272
Beilstein 1757502
Solubility Information Soluble in water (16g/L) (25°C)
Formula Weight 142.08
Percent Purity 97%
Chemical Name or Material 2,2,2-Trifluoroethyl acetate
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit