Learn More
Chemglass Life Sciences Funnel, 125mL, Separatory, Blank, Stopper Neck, Size # 22, Glassblowers
Supplier: Chemglass Life Sciences CG62403
Funnel, 125mL, Separatory, Blank, Stopper Neck, Size # 22, Glassblowers. Squibb shaped separatory blank complete with tooled and finished ground stopper neck top. Lower portion of blank is unfinished and requires a stopcock to be sealed. NOTE: SUPPLIED WITHOUT STOPPER.