Learn More
Electron Microscopy Sciences Critical Point Dryer 12 Specimen Holder
Supplier: Electron Microscopy Sciences 7019412
Machined from solid steel and brass/nickel plated. Incorporates a stainless steel mesh top and bottom to allow for fluid exchange. Comes with two sections; each section holds 6 specimens in 6 cavities cavity measures 7.5mm (D)x6mm(H)..