Learn More
Bioassay Systems QuantiChrom™ Β-glucosidase Assay Kit. For quantitative determination of β-glucosidase activity and evaluation of drug effects on its metabolism.
Supplier: Bioassay Systems DBGD100
For quantitative determination of β-glucosidase activity and evaluation of drug effects on its metabolism. Key Features: High sensitivity and wide linear range. Use 20 µL sample. The detection limit is 2 U/L, linear up to 250 U/L. Homogeneous and simple procedure. Simple "mix-and-measure" procedure allows reliable quantitation of β-glucosidase activity within 20 minutes. Robust and amenable to HTS. All reagents are compatible with high-throughput liquid handling instruments. Method: OD405nm. Samples: Biological. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 2 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.