Learn More
Bioassay Systems QuantiFluo™ Diamine Oxidase Assay Kit. For quantitative determination of diamine oxidase in biological samples (e.g. serum, plasma etc).
Supplier: Bioassay Systems QFDO100
For quantitative determination of diamine oxidase and evaluation of drug modulators. Key Features: Fast and sensitive. Use of 10 µL sample. Linear detection range 0.5 to 6 U/L for 30 min reaction at 25°C. Convenient. The procedure involves adding a single working reagent, and reading the fluorescence at 0 and 30 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. "Add-mix-read" type assay. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: FL530/585nm. Samples: Serum, Plasma. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: 0.5 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.