Learn More
Biotang Inc EGTA (0.5M, pH 8.0), autoclaved. 500 mM EGTA solution, pH 8.0. Prepared in 18.2 megohms water and autoclaved at 15 psi for 30 min on liquid cycle. Shipped at room temperature. Store at room temperature.
Supplier: Biotang Inc BM151
EGTA (0.5M, pH 8.0), autoclaved. 500 mM EGTA solution, pH 8.0. Prepared in 18.2 megohms water and autoclaved at 15 psi for 30 min on liquid cycle. Shipped at room temperature. Store at room temperature.