Thermo Scientific CO2 Incubators
Thermo Scientific CO2 Incubators
Featured Products
Culture Confidently
Thermo Scientific CO2 incubators let you culture with confidence — day after day and year after year. They offer innovative features like solutions to support your GMP and cleanroom needs, HEPA air filtration that surrounds cells with clean room-like air quality, and a choice of 100% pure copper or polished stainless-steel interior surfaces. Plus, the direct-heat units feature a high-temperature decontamination function to eliminate the need to handle or autoclave components.
Shop Thermo Scientific CO2 Incubators
thermo_scientific_logo
Cleanroom & Specialty CO2 Incubators
Designed for Compatibility and Control
Choose from CO2 incubators designed to support your cleanroom and other specialty needs, including GMP compatibility, high-volume culturing, and high contamination control.
heracell-160i-3d-22-0684
Heracell VIOS 160i/250i CR CO2 Incubators
Thermo Scientific™ Heracell™ Vios CR CO2 Incubators are certified cleanroom compatible. With active particle control and cleaning in mind, they also include a factory compliance acceptance package to help support your GMP documentation requirements.
View 3D Tour of the Heracell Vios 160i CR CO2 Incubator
View 3D Tour of the Heracell Vios 250i CR CO2 Incubator
Heracell VIOS CR
Thermo Scientific™ Heracell™ VIOS 160i/250i CR CO2 Incubators
Explore solutions for commercial and gene therapy manufacturing.
CO2 Incubator, 165 L, Electropolished Stainless Steel
CO2 Incubator, 165 L, Copper
CO2 Incubator, 255 L, Electropolished Stainless Steel
CO2 Incubator, 255 L, Copper
Forma Stericult CTS
Thermo Scientific™ Forma™ Steri-Cult™ CTS CO2 Incubators
Carry out high-volume culturing with a high level of contamination control with these stackable CO2 incubators, complete with documentation and validation to support GMP production needs.
CO2 Incubator, 232 L
CO2 Incubator, 323 L
Heracell VIOS 160i
Thermo Scientific™ Heracell™ VIOS 160i CO2 Incubators with Cell Locker System
Feature removable, protected chambers designed for improved culturing efficiency and security for sensitive cultures.
CO2 Incubator, 1-21% O2 Control, Stainless Steel, IR Sensor, 120V
CO2 Incubator, 1-21% O2 Control, Copper, IR Sensor, 120V
CO2 Incubator, Stainless Steel, IR Sensor, 120V
CO2 Incubator, Copper, IR Sensor, 120V
Tri-Gas CO2 Incubators
Variable Oxygen Control
Many cell cultures thrive best in CO2 incubators with controlled levels of oxygen. Select an O2 option to simulate physiological hypoxic environments (for stem cell and IVF applications) or choose to increase oxygen concentration for the ability to operate at hyperoxic levels.
Heracell VIOS
Thermo Scientific™ Heracell™ VIOS 160i/250i CO2 Incubators with O2 Control
The newest Thermo Scientific incubator series provides everything necessary for your most demanding and highly critical applications, combining the latest technology advancements in contamination control and uniform growth conditions with existing reliable features.
160i CO2 Incubator, Copper
160i CO2 Incubator, Stainless Steel
250i CO2 Incubator, Copper
250i CO2 Incubator, Stainless Steel
Heracell General Purpose
Thermo Scientific™ Heracell™ 150i/240i CO2 Incubators with O2 Control
Renowned for accuracy, uniformity, and quick recovery rates, Thermo Scientific Heracell 150i and 240i Direct Heat Incubators combine optimal culturing conditions with simplicity and ease of use.
150i CO2 Incubator, Stainless Steel
150i CO2 Incubator, Copper
240i CO2 Incubator, Stainless steel
240i CO2 Incubator, Copper
Forma Series
Thermo Scientific™ Forma™ Water-Jacketed CO2 Incubators
Benefit from the precise control, parameter recovery, dependable temperature stability, and HEPA cleanroom air purity of Series 3 and Series II incubators.
Series 3 CO2 Incubator, TC Sensor, 184 L
Series 3 CO2 Incubator, IR Sensor, 184 L
Series II CO2 Incubator, TC Sensor, 184 L
Series II CO2 Incubator, IR Sensor, 184 L
Direct Heat CO2 Incubators
Even Temperatures
Direct heat CO2 incubators feature efficient, high-performance heaters located on every chamber surface, providing even temperature distribution throughout the entire chamber.
foram-steri-cult-3d-22-0684
Forma Steri-Cult CO2 Incubators
Perform high-volume culturing with these stackable Thermo Scientific™ Forma™ Steri-Cult™ CO2 Incubators. They offer CO2 sensors, humidity control, HEPA filtration, and a decontamination cycle.
Shop Forma Steri-Cult CO2 Incubators
View 3D Tour of the Forma Steri-Cult CTS CO2 Incubator
Heracell
Thermo Scientific™ Heracell™ VIOS CO2 Incubators
Choose the newest Thermo Scientific incubator series, which combines the latest technology advancements in contamination control and uniform growth conditions with existing reliable features.
160i CO2 Incubator, 165 L, Copper
160i CO2 Incubator, 165 L, Electropolished Stainless Steel
250i CO2 Incubator, 255 L, Electropolished Stainless Steel
250i CO2 Incubator, 255 L, Copper
Forma Direct Heat
Thermo Scientific™ Heracell™ VIOS 150i and 240i CO2 Incubators
Renowned for accuracy, uniformity, and quick recovery rates, Thermo Scientific Heracell 150i and 240i Direct Heat Incubators combine optimal culturing conditions with simplicity and ease of use.
150i CO2 Incubator, 150 L, Copper, General Purpose
150i CO2 Incubator, 150 L, Stainless Steel, General Purpose
240i CO2 Incubator, 240 L, Electropolished Stainless Steel, General Purpose
240i CO2 Incubator, 240 L, Copper, General Purpose
Reach/Midi
Thermo Scientific™ Reach/Midi CO2 Incubators
Choose Reach and Midi CO2 Incubators for your large- and small-capacity needs.
Large-Capacity Reach-In CO2 Incubator, 821 L
Midi CO2 Incubator, 40 L
Water-Jacketed CO2 Incubators
Dual Insulation
Water-jacketed CO2 incubators offer unique triple-wall construction, providing temperature stability with dual layers of water and insulation.
Forma Series 4
Thermo Scientific™ Forma™ Series II CO2 Incubators
Forma Series II Water Jacketed CO2 Incubators are designed to deliver dependable thermal protection and quick recovery from swings in ambient temperature and power variations.
CO2 Incubator, 184 L, TC Sensor
CO2 Incubator, 184 L, IR Sensor
Forma Series 3
Thermo Scientific™ Forma™ Series 3 CO2 Incubators
Forma Series 3 Water Jacketed CO2 Incubators deliver the outstanding performance of the Series II with a touchscreen iCAN interface.
CO2 Incubator, 184 L, TC Sensor
CO2 Incubator, 184 L, IR Sensor
Resources
More
Special Offers