Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zonisamide, USP, 98-101%, Spectrum™ Chemical

Click to view available options
Quantity:
5 g
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Zonisamide, USP
Specifications
CAS Max % | 1 |
Ignition Residue | 0.002 |
Packaging | Amber Glass Bottle |
Quantity | 5 g |
Percent Purity | 98 to 101% |
Grade | USP |
Water | 0.01 |