Learn More
Thermo Scientific™ Zinc PVA Fixative Single Vial
Collect, preserve and permanently stain intestinal parasites with a zinc-based fixative, part of system which comes with industry standard color-coding and sporked caps.
Supplier: Thermo Scientific™ R21927
Description
Collect, preserve and permanently stain intestinal parasites with zinc-based Thermo Scientfic™ Zinc PVA Fixative Single Vial, part of a complete collection of quality fecal transports which come with industry standard color-coding and easy-to-use sporked caps.
- Broad range
- Long shelf life with industry standard color-coded caps and labels
- Easy to use spork cap for safe collection
- Convenient packaging, including bulk kits
- Multilingual patient collection instructions
Note:
Zinc PVA is copper-based.
Specifications
Zinc PVA Fixative | |
15 mL | |
120 Vials/Pk. |
Zinc PVA Fixative Single Vial | |
Single vial. Packaged in partitioned boxes. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.