Learn More
ZeptoMetrix SIV Western Blot Assay Kit
For the in vitro detection of antibodies to SIV in serum or plasma
Specifications
Product Type | Simian Immunodeficiency Virus (SIV) Western Blot Kit |
---|---|
Content And Storage | 2°C to 8°C |
Includes: SIV Nitrocellulose Strips (10 or 30 strips), Non-Reactive Control (1 vial, 100μL), Reactive Control (1 vial, 100μL), Blotting Buffer (1 bottle, 125mL), 20X Wash Buffer (1 bottle, 100mL), Conjugate (1 vial, 100μL), Substrate (1 bottle, 75mL), Blotting Powder (1 packet, 10g) and Incubation Trays
Requires: Rocking platform, Aspirator with disinfectant trap, Disposable gloves, Calibrated adjustable micropipettes, Graduated cylinders and assorted beakers, Timer, 1% sodium hypochlorite as disinfectant. May be prepared from household bleach, Distilled or deionized water and Tweezers or forceps
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Quantity | Price | Quantity & Availability | ||||
22-157-381
|
ZeptoMetrix
0801501 |
Simian Immunodeficiency Virus (SIV) Western Blot Kit | 30 Strips |
Each for $1,914.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-380
|
ZeptoMetrix
0801500 |
Simian Immunodeficiency Virus (SIV) Western Blot Kit | 10 Strips |
Each for $772.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
22-157-382
|
ZeptoMetrix
0801502 |
Simian Immunodeficiency Virus (SIV) Western Blot Kit | 10 Strips |
Each for $446.00
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- The assay utilizes nitrocellulose strips that contain SIVMAC viral proteins resulting from electrophoresis and electroblotting of detergent disrupted virus particles
- A serum control band is included on each strip to minimize the risk of false negative results due to operational error
Specifications
Simian Immunodeficiency Virus (SIV) Western Blot Kit | |
2°C to 8°C |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.